Learn More
Memmert™ Special Flat Stainless Steel Cover with Openings and Concentric Ring Sets
Flat Stainless steel cover with openings and concentric ring sets
Marca: Memmert™ WTB15B48808
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Only in combination with CDP115/ CDP115X2/ CPM650
Specifica
WTB15 | |
Special flat stainless steel cover with openings and concentric ring sets | |
Stainless Steel |