Learn More
Biochrom Spectrophotometer Sipper
Spectrophotometer Sipper complete with UV quartz cell and all tubing. For use with the Ultrospec 7500 and 2100 Pro Spectrophotometer.
Marca: Biochrom 80-2112-15
Dettagli aggiuntivi : Peso : 1.14900kg
Descrizione
- For use with the Ultrospec 2100 Pro spectrophotometer
- Requires single cell holder 80-2106-05 or 80-2106-13
Specifica
Sipper System | |
Ultrospec 7500 and 2100 Pro Spectrophotometer | |
UV quartz cell and tubing | |
Single Cell Holder 80-2106-05 or 80-2106-13 |