Learn More
MP Biomedicals™ SPINeasy™ DNA Kit for Blood
Fast and efficient DNA isolation from blood and other biofluids in less than 30 minutes. Convenient spin-column technology with no organic solvents. High DNA yield and purity, free of common contaminants. DNA ready for downstream PCR or NGS applications.
Marca: MP Biomedicals™ 116552050
Dettagli aggiuntivi : Peso : 0.32120kg
Descrizione
- SPINeasy DNA Kit for Blood is a high-performance genomic DNA (gDNA) extraction kit based on silica-membrane spin-column technology.
- This kit enables quick isolation of gDNA from whole blood preserved in different anticoagulants (EDTA, heparin, and sodium citrate), typically in less than 30 minutes.
- It also allows easy gDNA isolation from coagulated blood when using the FastPrep-24 5G homogenizer with lysing matrix A or D tubes (recommended for soft tissues, and not included with this kit).
- The use of our specially formulated Lysis Buffer BL enables highly efficient lysis of various liquid samples including plasma, serum, saliva, and cell culture medium.
- Column S1 and kit buffers (provided in the kit) are designed to deliver gDNA extracts of high yield and purity, compatible with downstream molecular applications such as PCR and NGS.
Specifica
Store at Room Temperature, Dry Environment | |
Lysis Buffer BL, Wash Buffer BW1, Wash Buffer BW2, Elution Buffer EB, Column S, 2.0 mL collection tubes,1.5 mL microcentrifuge tubes, Proteinase K, Proteinase K Buffer | |
200 µL of Human Blood, Serum, Plasma, Saliva, or Cell Culture Media |
Spin Column | |
50 Preps |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.