missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17976-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody specifically recognizes the 54 kDa SRP54 protein.
Binds to the signal sequence of presecretory protein when they emerge from the ribosomes and transfers them to TRAM (translocating chain-associating membrane protein).Specifica
SRP54 | |
Polyclonal | |
Unconjugated | |
SRP54 | |
SRP54 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116650, 24067, 6729 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14576, P61011, Q6AYB5 | |
SRP54 | |
SRP54 Fusion Protein Ag12166 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |