Learn More
SSC5D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559819
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: TCTGYTDYDD YPPWTWDPTS REDLAKGTTT AGVPGHTLPW RTTRRPGSSS PAIRRLPDTG SKDGYKLPWT WDTPSGRGLA EGTPTAGKLG PTL Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 61%.
Binds to extracellular matrix proteins. Binds to pathogen-associated molecular patterns (PAMPs) present on the cell walls of Gram-positive and Gram-negative bacteria and fungi, behaving as a pattern recognition receptor (PRR). Induces bacterial and fungal aggregation and subsequent inhibition of PAMP-induced cytokine release. Does not possess intrinsic bactericidal activity. May play a role in the innate defense and homeostasis of certain epithelial surfaces.Specifica
SSC5D | |
Polyclonal | |
Unconjugated | |
SSC5D | |
S5D-SRCRB; scavenger receptor cysteine rich domain containing (5 domains); scavenger receptor cysteine rich family member with 5 domains; scavenger receptor cysteine rich family, 5 domains; scavenger receptor cysteine-rich glycoprotein; soluble scavenger protein with 5 SRCR domains; Soluble scavenger receptor cysteine-rich domain-containing protein SSC5D; soluble scavenger with 5 domains; SSC5D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284297 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.13 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1L4H1 | |
SSC5D | |
Recombinant protein corresponding to Human SSC5D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only