missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11945-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The gene for this product maps to a region on chromosome 7 identified as an autism-susceptibility locus. Mutation screening of the entire coding region in autistic individuals failed to identify phenotype-specific variants, suggesting that coding mutations for this gene are unlikely to be involved in the etiology of autism. The function of this gene product has not been determined. Transcript variants encoding different isoforms of this protein have been described.Specifica
ST7 | |
Polyclonal | |
Unconjugated | |
ST7 | |
ETS7q, FAM4A, FAM4A1, HELG, Protein FAM4A1, Protein HELG, RAY1, SEN4, ST7, TSG7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64213, 7982 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99M96, Q9NRC1 | |
St7 | |
ST7 Fusion Protein Ag2547 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |