Learn More
De Buyer™ Placca di cottura in acciaio inox
Use this De Buyer™ Stainless-Steel Culinary Tray with straight edges for baking, presentation and food storage. The tray is robust, sturdy and undeformable and has a mirror-like finish.
Marca: De Buyer™ 332065
Dettagli aggiuntivi : Peso : 2.87000kg
Descrizione
- 1mm thickness
- Mirror-like finish
- Can be used with a silicone mat or baking paper for cold forming (easier mould release) and cooking
- Provides support for de Buyer metal moulds, rings and frames
- Resists very low temperatures and humidity
Specifica
400 x 300 mm | |
20 mm | |
Stainless Steel | |
300 mm | |
Rectangular |
400 x 300 x 20 mm | |
400 mm | |
Tray | |
1 Each | |
1 mm |