Learn More
De Buyer™ Placca di cottura in acciaio inox
Use this De Buyer™ Stainless-Steel Culinary Tray with straight edges for baking, presentation and food storage. The tray is robust, sturdy and undeformable and has a mirror-like finish.
Marca: De Buyer™ 332032
Dettagli aggiuntivi : Peso : 0.67000kg
Descrizione
- 1mm thickness
- Mirror-like finish
- Can be used with a silicone mat or baking paper for cold forming (easier mould release) and cooking
- Provides support for de Buyer metal moulds, rings and frames
- Resists very low temperatures and humidity
Specifica
325 x 265 mm | |
20 mm | |
Acciaio inox | |
265 mm | |
Rectangular |
325 x 265 x 20 mm | |
325 mm | |
Vassoio di cottura | |
1 ciascuno | |
1 mm |