Learn More
Masterflex™ Stainless Steel Barbed Fitting NPT Male Pipe Adapter
Designed for quick and easy assembly without tools
Marca: Masterflex™ 31208-86
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Available in a variety of materials and configurations to meet your need
- Adapts from a NPT male thread to a hose barb
Specifica
Barbed Fitting NPT Male Pipe Adapter | |
Stainless Steel | |
3/8 in. | |
3/8 in. NPT(M) | |
3/8 in. NPT(M) |
3/8 NPT(M) x 3/8 in. Tubing ID | |
1 7/32 in. | |
3/8 in. | |
3/8 in. | |
11/16 in. |