Learn More
Ohaus™ Stainless Steel Calibration Weight Weight - OIML E1 Wire, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E1 Tolerances
Marca: Ohaus™ 30517968
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
Specifica
NVLAP Accredited Certificate | |
50 mg | |
Protective Case |
Stainless Steel (316) | |
0.004 mg |