Learn More
Smeg™ Stainless Steel Instrument Basket
Designed for use with Smeg™ glassware washers. Smeg Stainless Steel Instrument Baskets are suitable for medium-sized instruments.
Marca: Smeg™ CSK1/3
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Baskets with stainless steel handles, suitable for medium-sized instruments
- Mesh openings of 5 x 5mm
- Dimensions: 225 x 450 x 50mm
Specifica
435 x 160 x 50mm |