Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50mmH, with Perforated Plates with Slotted Holes
Marca: RETSCH 60.180.001800
Dettagli aggiuntivi : Peso : 1.00000kg
Specifica
Stainless Steel | |
Stainless Steel | |
1 Each | |
Test Sieve | |
Certificate indicating Compliance |
For separation, fractioning, particle size determination | |
200 mm | |
1.8mm | |
ISO 5223 | |
50mm |