missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stat1 (pY701) Mouse, BV421, Clone: 4a, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566238
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Stat1 (pY701) Monoclonal antibody specifically detects Stat1 (pY701) in Human, Mouse, Rat samples. It is validated for Flow cytometry (Intracellular Staining)) .Specifica
Stat1 (pY701) | |
Monoclonal | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
4a | |
Vol. Per test: 5μL | |
Signal Transducer and Activator of Transcription-1; STAT91; ISGF-3; CANDF7 | |
Phosphorylated Human Stat1 Peptide | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2a |
For Reseach Use Only.