missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5 alpha Monoclonal Antibody (251610), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marca: Thermo Scientific MA523571
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody does not cross-react with STAT5b.
STAT5 alpha Monoclonal antibody specifically detects STAT5 alpha in Human samples. It is validated for Flow Cytometry.Specifica
STAT5 alpha | |
Monoclonal | |
FITC | |
STAT5A | |
AA959963; mammary gland factor; mammary gland factor STAT5A; mammary gland factor; transcription factor; Mgf; Mpf; signal transducer and activator of transcription 5; signal transducer and activator of transcription 5a; signal transducer and activator of transcription factor 5A; STAT; STAT5; STAT5A; STAT5A, Mammary Gland Factor; STAT5A/MGF; STAT5B; STATA5 | |
Mouse | |
Protein A/G | |
RUO | |
6776 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
251610 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P42229 | |
STAT5A | |
Human STAT5a synthetic peptide SLDSRLSPPAGLFTSARGSLS | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |