missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12071-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human STAT5B.
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. This gene was found to fuse to retinoic acid receptor-alpha (RARA) gene in a small subset of acute promyelocytic leukemias (APLL). The dysregulation of the signaling pathways mediated by this protein may be the cause of the APLL.Specifica
STAT5B | |
Polyclonal | |
Unconjugated | |
STAT5B | |
STAT5, STAT5B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20851, 25126, 6777 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P42232, P51692, P52632 | |
STAT5B | |
STAT5B Fusion Protein Ag2709 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |