Learn More
Abeba™ Static Control 31474 Shoes
ESD dissipative and Sanitized™ treated.
Marca: Abeba™ 31474/41
Dettagli aggiuntivi : Peso : 1.30000kg
Descrizione
- CE EN ISO 20345:2011 S3 SRA
- Smooth leather
- Black in ATEX design
- Composite toe cap
- PU midsole
- Metalfree penetration resistance
- Slip resistance special rubber outsole sole
- Replaceable acc Wave insole (article no. 3553)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Specifica
CE EN ISO 20345:2011 S3 SRA | |
Leather | |
Slip-resistant | |
31474 |
Black | |
41 | |
Rubber | |
Composite |