missing translation for 'onlineSavingsMsg'
Learn More

STK3 Mouse anti-Human, Mouse, Rat, Clone: 1C11A4, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66637-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898695

  • 325.38€ / 150µL

Spedizione stimata: 10-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
Specifica

Specifica

STK3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O54748, Q13188, Q9JI10
Stk3
STK3 Fusion Protein Ag2741
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG2a
Western Blot
1C11A4
Unconjugated
STK3
KRS1, MST 2, MST2, STE20 like kinase MST2, STK3
Mouse
Protein A
RUO
56274, 65189, 6788
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali