Learn More
Gosselin™ Straight Specimen Containers with Red Cap
CORNING GOSSELIN™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
Marca: Gosselin™ TP35C-052
Dettagli aggiuntivi : Peso : 4.96000kg
Descrizione
- Clear and break-resistant polypropylene (PP)
- Available in different sizes and quantities
- Color-coded screw caps
Specifica
Specimen Container | |
Polypropylene | |
Sterile | |
Red | |
Clear | |
600/Cs. |
Yes | |
Straight | |
60 mL | |
Screw Cap | |
No Label | |
30/Tray |