Learn More
STXBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 20562-1-AP-20UL
 vedi altre versioni di questo prodotto
Codice prodotto. 16890115
  • 145.00€ / 20µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
The antibody is specific to STXBP1.
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
Specifica
Specifica
STXBP1
Polyclonal
Unconjugated
STXBP1
EIEE4, MUNC18 1, N Sec1, p67, Protein unc 18 homolog 1, Protein unc 18 homolog A, RBSEC1, STXBP1, syntaxin binding protein 1, Unc 18A, UNC18, Unc18 1, UNC18A
Rabbit
Antigen Affinity Chromatography
RUO
20910, 25558, 6812
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08599, P61764, P61765
STXBP1
Peptide
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.