Learn More
Superoxide Dismutase 1 (SOD1) (Antioxidant Enzyme) Mouse, Clone: SOD1/2089, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 6647-MSM1-P0
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| Superoxide Dismutase 1 (SOD1) (Antioxidant Enzyme) | |
| Cytoplasmic | |
| SOD1/2089 | |
| Unconjugated | |
| Mouse | |
| 23kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 6647 | |
| IgG |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human SOD1 protein | |
| HeLa or Jurkat Cells. Breast or Ovarian carcinoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |