Learn More
Cytiva (formerly Pall Lab) Supor™ Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
Marca: Cytiva (formerly Pall Lab) 61854
Dettagli aggiuntivi : Peso : 0.24700kg
Descrizione
- Optimized for biological, pharmaceutical and sterilizing filtration requirements
Specifica
PES | |
Sterile | |
140 μm | |
47 mm |
0.45 μm | |
100/Pk. | |
Gridded |