Learn More
SVL™ Sealing Ring for Butt Joint
Manufactured from silicone rubber
Marca: SVL 701-01
61.09 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Silicone rubber and sheathed with PTFE for protection against chemical attack
- Internal diameter of ring corresponds to the bore of the SVL glass screwthread so that flow is not restricted
Specifica
Joint size: 15 | |
Butt joints | |
Sealing Ring | |
Silicone Rubber sheathed with PTFE |