Learn More
Notrax Swisslon XT™ Premium Quality Barrier Matting, Charcoal
Notrax Swisslon XT™ Premium quality barrier matting designed to remove walked-in moisture and finer dirt particles in large indoor entrance areas, including wall-to-wall applications.
Marke: Notrax 380S0035CH
Weitere Details : Peso : 4.90000kg
Beschreibung
- Premium quality barrier matting designed to remove walked-in moisture and finer dirt particles in large indoor entrance areas, including wall-to-wall applications
- Our redesigned classic barrier mat, known for its softer yarns, has been enhanced with a mono-filament to offer additional scraping and provide excellent cleaning and absorbing
- The new tri-color grit pattern provides outstanding dirt-hiding characteristics
- Great ability to retain moisture (<58 liters per m²) and scrape debris from foot traffic while maintaining its great looking appearance for the longest time
- A dense heavyweight vinyl backing ensures longevity
Spezifikation
59 in. | |
Industrial and Commercial Matting | |
90 cm |
Charcoal | |
Polyamide 6.0 Yarns | |
150 cm |