missing translation for 'onlineSavingsMsg'
Learn More

Synaptophysin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5B3F2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60191-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897175

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.
Specifica

Specifica

Synaptophysin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07825, P08247, Q62277
SYP
Synaptophysin Fusion Protein Ag11803
150 μL
Primary
Rat, Porcine, Mouse, Human
Antibody
IgG3
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
5B3F2
Unconjugated
SYP
synaptophysin, SYP
Mouse
Protein G
RUO
100512029, 20977, 24804, 6855
-20°C
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali