Learn More
Trajan Scientific and Medical™ Syringe Hitachi LC autosampler M10 x 1 thread 500μL SGE
For HPLC Applications that provide accurate and reproducible injections
Marca: Trajan Scientific and Medical™ 007660
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Hitachi instruments
Specifica
PTFE (Tip) | |
500 μL | |
Gas Tight |
HPLC | |
Hitachi LC Autosamplers |