Learn More
TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results Epredia™ TACE (TNF-α converting enzyme)/ADAM17 Ab-1, Rabbit Polyclonal Antibody.
Marca: Epredia RB-1660-P
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
TACE (TNF-α converting enzyme) protein is a membrane bound matalloprotease from ADAM (a disintegrin and metalloprotease) family of proteins. It cleaves 26kDa type II membrane-bound precursor of TNF-α into 17kDA mature form which is secreted. TACE is also known to process other membrane proteins including TNF receptor, TGFα, L-selectin and amyloid protein precursor.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide from the C-terminus of human TACE protein
Molecular Weight: 80-130kDa (corresponding to mature, precursor and glycosylated forms)
Positive Control: HeLa cells
Cellular Localization: Membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifica
TACE (TNF-alpha converting enzyme)/ADAM17 Ab-1 | |
Polyclonal | |
1mg/mL | |
Rabbit | |
1 mL | |
Proteases And Inhibitors | |
Human, Mouse | |
IgG |
Western Blot | |
Polyclonal | |
Unconjugated | |
A synthetic peptide from the C-terminus of human TACE protein | |
RUO | |
Primary | |
Concentrated |