missing translation for 'onlineSavingsMsg'
Learn More

TCN2 Mouse anti-Human, Clone: 2G6D2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66377-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898195

  • 121.38€ / 20µL

Spedizione stimata: 10-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Transcobalamin I (TCI) and Transcobalamin II (TCII) are secreted proteins belonging to the eukaryotic cobalamin transport proteins family and also to the vitamin B12-binding protein family. The genes encoding these proteins map to chromosome 11q11-q12 and 22q12.2, respectively. Transcobalamin I is a constituent of secondary granules in neutrophils, while Transcobalamin II binds cobalamin and mediates its transport into cells. These plasma proteins are expressed in various tissues and secretions.
Specifica

Specifica

TCN2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20062
TCN2
TCN2 Fusion Protein Ag2825
20 μL
Primary
Human
Antibody
IgG2b
Western Blot
2G6D2
Unconjugated
TCN2
TC 2, TC II, TC2, TCII, TCN2, Transcobalamin 2, Transcobalamin II
Mouse
Protein A
RUO
6948
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali