missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR Vβ5.1 Mouse anti-Human , PE, Clone: LC4, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 567042
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
TCR Vβ5.1 Monoclonal antibody specifically detects TCR Vβ5.1 in Human samples. It is validated for Flow CytometrySpecifica
TCR Vβ5.1 | |
Monoclonal | |
PE | |
TCR V beta 5.1; TCR Vb5.1; TRBV5-1; TRBV51 | |
Human T acute lymphoblastic leukemia Cell Line | |
RUO | |
Human | |
Mouse BALB/c IgG1 κ |
Flow Cytometry | |
LC4 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |