missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEM7/PLXDC1 Antibody (197C193 (IM193)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
Marca: Novus Biologicals NB100-56557
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TEM7/PLXDC1 Monoclonal specifically detects TEM7/PLXDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifica
| TEM7/PLXDC1 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Meng et al (2007)), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (Nanda et al (2004)), Immunohistochemistry-Paraffin 2-5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (Lee et al (2006)) | |
| FLJ45632, plexin domain containing 1,2410003I07Rik, plexin domain-containing protein 1, TEM3FLJ36270, TEM7DKFZp686F0937, Tumor endothelial marker 3, Tumor endothelial marker 7 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 197C193 (IM193) | |
| Unconjugated | |
| Q8IUK5 | |
| PLXDC1 | |
| Amino acids 409-425 (LQNNLSPKTKGTPVHLG) of human TEM7 were used to develop this monoclonal antibody. | |
| 0.1 mg | |
| Angiogenesis | |
| 57125 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto