Learn More
tert-Butyl diethylphosphonoacetate, 95%, Thermo Scientific Chemicals
Descrizione
tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,β-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,ß-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifica
Materiale o nome chimico | tert-Butyl diethylphosphonoacetate |
Densità | 1.074 |
Punti di ebollizione | 100°C to 103°C (1.5 mmHg) |
Punto d'infiammabilità | >110°C (230°F) |
Indice di rifrazione | 1.431 |
Quantità | 1 g |
Beilstein | 2050126 |
Informazioni di solubilità | Not miscible or difficult to mix in water. |
Peso formulazione | 252.25 |
Percent Purity | 95% |
RUO – Research Use Only