Learn More
Smeg™ Test Tube Basket, 200 mm Max. Height
Contains 3 removable compartments and lid.
Marca:  Smeg™ CP222
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11995554
  • 395.00€ / Pezzo
Spedizione stimata: 10-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Additional accessory for the standard baskets CS1-1 and CS2 and the trolley LM20 for washing analysis test tubes.
  • Contains 3 removable compartments and lid
  • Takes up one quarter of the space of a standard basket
Specifica
Specifica
160 Standard Test Tubes
Basket with Removable Compartments and Lid
7.87 in.
200 mm