Learn More
Biochrom Test Tube Cell Holder for Novaspec III+ and Novaspec Pro Spectrophotometers
Test Tube Cell Holder for Novaspec III+ and Novaspec Pro Spectrophotometers is spring loaded for 10 to 18 mm diameter test tubes.
Marca: Biochrom 80-3007-12
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Test tube cell holder
- For use with Biochrom Novaspec III+ and Novaspec Pro Spectrophotometers
Specifica
Novaspec III+ and Novaspec Pro Spectrophotometers | |
Reusable | |
10 to 18 mm Diameter Test Tubes |
Test Tube Cell Holder | |
Biochrom Novaspec III+ and Novaspec Pro Spectrophotometers | |
Cylindrical |