Learn More
Edmund Bühler GmbH™ Test Tube Rack
Constructed of Stainless steel. Edmund Bühler GmbH™ Test Tube Rack for use with hinged foot on universal tray or with clamping strip on rack system.
Marca: Edmund Bühler GmbH™ 0052 201
Dettagli aggiuntivi : Peso : 0.05000kg
Descrizione
- Stainless steel construction
- Dimensions: 289 x 100mm
- Attach to hinged foot for angle variation (order separately)
- For use with universal platform shakers
Specifica
Tube Rack | |
Tubes (30 mm) | |
14 | |
30 mm |
Metal | |
Stainless Steel | |
Non-sterile | |
30 mm |