missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFII I Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10499-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.Specifica
TFII I | |
Polyclonal | |
Unconjugated | |
GTF2I | |
BAP 135, BAP135, BTK associated protein 135, BTKAP1, DIWS, GTF2I, GTFII I, IB291, SPIN, SRF Phox1 interacting protein, TFII I, WBS, WBSCR6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14886, 2969, 353256 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78347, Q5U2Y1, Q9ESZ8 | |
Gtf2i | |
TFII I Fusion Protein Ag0773 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |