Learn More
TGF-beta Antibody (TGFB/7230), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-23694-20ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TGF-beta Monoclonal antibody specifically detects TGF-beta in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifica
| TGF-beta | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Recombinant fragment (around aa200-390) of human TGF-beta protein | |
| 20 μg | |
| Cytokine Research, Extracellular Matrix | |
| 7040 | |
| Store at 4°C. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| TGFB/7230 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |