Learn More
THAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558929
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: KSNISSQQVL LEHSYAFRNP MEAKKRIIKL EKEIASLRRK MKTCLQKERR ATRRWIKATC LVKNLEANSV LPKGTSEHML PTALSSLPLE D Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.
THAP2, also named as THAP domain-containing protein 2, is a 228 amino acid protein, which contains one THAP-type zinc finger. Members of the THAP (thanatos-associated protein) family of proteins contain a well conserved DNA-binding domain known as the THAP-type zinc finger motif. Proteins containing the THAP-type zinc finger motif are commonly involved in transcriptional regulation, cell-cycle control, apoptosis and chromatin modification.Specifica
THAP2 | |
Polyclonal | |
Unconjugated | |
THAP2 | |
2900040O07Rik; 9030625G08Rik; AI450385; AI649097; THAP domain containing 2; THAP domain containing, apoptosis associated protein 2; THAP domain-containing protein 2; Thap2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83591 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0W7 | |
THAP2 | |
Recombinant protein corresponding to Human THAP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only