Learn More
Thermo Scientific™ Rebinding Buffer for MagJET Blood RNA Kit (concentrated)
Thermo Scientific™ MagJET™ Rebinding Buffer is a component of the MagJET™ Whole Blood RNA Kit (Cat. No. K2751) and may be purchased separately when additional solution is needed. It is provided in 20 mL bottles.
Marca: Thermo Scientific™ R2401
55.34 EUR valido fino al 2024-12-31
Usa il codice promo "21969" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.07000kg
Descrizione
The Thermo Scientific™ MagJET™ Whole Blood RNA Kit is designed for fast and efficient purification of total RNA from fresh or frozen whole blood treated with EDTA or citrate, blood fractions, bone marrow, or samples stabilized with PAXgene™ or Tempus™. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifica
Rebinding Buffer | |
MagJET | |
20 mL |
For Research Use Only. Not for use in diagnostic procedures.