Learn More
Thermo Scientific™ High Purity Two Stage Brass Gas Regulators
Recommended for non-corrosive analytical and process applications requiring precise, stable, delivery pressure control
Marca: Thermo Scientific™ 60180-908
Dettagli aggiuntivi : Peso : 4.50000kg
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Descrizione
- For non-corrosive analytical and process applications
- Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
- Specially designed, convoluted, stainless steel diaphragm provides goodregulating performance and maximum purity integrity without the needfor a soft seal
- Capable of withstanding an internal vacuum
- All regulators supplied with 1/8 in. stainless steel output adaptor
- Built in check valve allows for cylinder change without contaminating the gas line
Specifica
0 to 200 psig | |
DIN 1 with Triple Filter |