missing translation for 'onlineSavingsMsg'
Learn More

TIAF1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 20328-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899844

  • 140.00€ / 20µL
Spedizione stimata: 16-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

TIAF-1 (TGFB1-induced anti-apoptotic factor 1), also known as MYO18A, is a TGFB1-induced anti-apoptotic factor that functions to inhibit the cytotoxic effects of TNF-alpha, TRADD and FADD. It localizes to the nucleus and significantly up-regulated in activated helper T lymphocytes (TH2) in chronic kidney and liver allograft rejection. Suppression of TIAF1 expression by siRNA prevents UV irradiation-mediated p53 phosphorylation and nuclear translocation. TIAF1 is essential in the p53- and WOX1-mediated cell death and shows an aggregation-dependent control of tumor progression and metastasis, and regulation of cell death.
Specifica

Specifica

TIAF1
Polyclonal
Unconjugated
TIAF1
MAJN, MYO18A, MYSPDZ, SPR210, TIAF1
Rabbit
Antigen Affinity Chromatography
RUO
9220
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95411
TIAF1
TIAF1 Fusion Protein Ag14023
20 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali