Learn More
Lovibond™ Tintometer™ Acidifying PT Rapid Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca: Lovibond™ 515491BT
Dettagli aggiuntivi : Peso : 0.30000kg
Descrizione
- Acidifying PT Rapid Tablets
- Available in packs of 100 and 250
Specifica
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Acidifying PT Rapid Tablets |