Learn More
Lovibond™ Tintometer™ Molybdate No. 2 HR Tablet
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca: Lovibond™ 513070BT
Dettagli aggiuntivi : Peso : 0.30000kg
Descrizione
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Range: 1 to 50mg/L
- Parameter: Molybdate LR, molybdate, HR molybdate
- Method: Thioglycolate
- For use in our test kits, colorimeters and spectrophotometers
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifica
Photometry Tablet Reagents | |
100 Tablets | |
Molybdate No. 2 HR Tablet |
Test kits, colorimeters and spectrophotometers | |
1 to 50mg/L |
Sicurezza e movimentazione
- Attenzione
Categoria di pericolo
- Lesione oculare grave/irritazione oculare Categoria 2
- Sensibilizzazione della pelle Categoria 1
Indicazioni di pericolo
- H319-Provoca grave irritazione oculare.
- H317-Può provocare una reazione allergica cutanea.
Consigli di prudenza
- P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
- P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
- P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.
- P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
- P333+P313-In caso di irritazione o eruzione della pelle: consultare un medico.
- P337+P313-Se l'irritazione degli occhi persiste, consultare un medico.
Informazioni supplementari
- MIXTURE LIST-Contiene: calcium thioglycolate trihydrate