Learn More
Lovibond™ Tintometer™ Thymol Blue Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca: Lovibond™ 511651BT
Dettagli aggiuntivi : Peso : 0.30000kg
Descrizione
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Thymol Blue Tablets
- Available in packs of 100 and 250
Specifica
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
250 Tablets | |
Thymol Blue Tablets |