missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM6SF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA564257
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: PTDACFVYIY QYEPYLRDPV AYPKVQ Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 88%.
TM6SF2 is a protein coding gene.Specifica
TM6SF2 | |
Polyclonal | |
Unconjugated | |
TM6SF2 | |
TM6SF2; Transmembrane 6 superfamily member 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53345 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BZW4 | |
TM6SF2 | |
Recombinant protein corresponding to Human TM6SF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only