Learn More
TM9SF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552176
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: LFVNRLDSVE SVLPYEYTAF DFCQASEGKR PSENLGQVLF GERIEPSPYK FTFNKKETCK LVCTKTYHTE KAEDKQKLEF LKKSMLLNYQ HHWIVDNMPV TWCYDVEDGQ RF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
In the intracellular compartments, may function as a channel or small molecule transporter.Specifica
TM9SF2 | |
Polyclonal | |
Unconjugated | |
TM9SF2 | |
76 kDa membrane protein; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 2; P76; TM9SF2; Transmembrane 9 superfamily member 2; transmembrane protein 9 superfamily member 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9375 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99805 | |
TM9SF2 | |
Recombinant protein corresponding to Human TM9SF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only