Learn More
TMEM120B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553122
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: THRIYKQKLE ELAALQTLCS SSISKQKKHL KDLKLTLQRC KRHASREEAE LVQQMAANIK ERQDVFFDME AYLPKKNGLY LNLVLGNVNV TLLSNQAKFA YKD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
TMEM120B also named as transmembrane protein 120B is 339 amino acid multi-pass membrane protein, which belongs to the TMEM120 family and localizes in membrane. The function of TMEM120B is still non-known.Specifica
TMEM120B | |
Polyclonal | |
Unconjugated | |
TMEM120B | |
Tmem120b; Transmembrane protein 120B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144404 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A0PK00 | |
TMEM120B | |
Recombinant protein corresponding to Human TMEM120B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only