missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM39B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559110
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: TSVRSRTRSS SGTGLSSPPL ATQTVVPLQH CKIPELPVQA SIL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
TMEM39B Polyclonal antibody specifically detects TMEM39B in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
TMEM39B | |
Polyclonal | |
Unconjugated | |
TMEM39B | |
TMEM39B; Transmembrane protein 39B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55116 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZU3 | |
TMEM39B | |
Recombinant protein corresponding to Human TMEM39B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only