Learn More
TNC/Tenascin-C Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27789-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The TNC/Tenascin-C antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TNC/Tenascin-C. This antibody recognizes human antigen. The TNC/Tenascin-C antibody has been validated for the following applications: WB,ELISA analysis.
Specifica
| TNC/Tenascin-C | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 3371 | |
| Liquid |
| Western Blot, ELISA | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human |