Learn More
TNFAIP2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 25649-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00020kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17251376
  • 121.38€ / 20µL
Spedizione stimata: 30-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
TNFAIP2, also named as B94, belongs to the SEC6 family. TNFAIP2 may play a role as a mediator of inflammation and angiogenesis. TNFAIP2 is differentially expressed in development and capillary tube-like formation in vitro. It is induced by TNFα and other proinflammatory factors.
Specifica
Specifica
TNFAIP2
Polyclonal
Unconjugated
20 μL
7127
Liquid
ELISA, Western Blot
700 μg/mL