Learn More
TNFAIP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23456-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody, which is a rabbit polyclonal antibody raised against 350 aa of C-terminus of human TNFAIP3, can recognize full-length A20 (∽90 kDa) and cleaved form (∽37-40 kDa).
This gene was identified as a gene whose expression is rapidly induced by the tumor necrosis factor (TNF). The protein encoded by this gene is a zinc finger protein, and has been shown to inhibit NF-kappa B activation as well as TNF-mediated apoptosis. Knockout studies of a similar gene in mice suggested that this gene is critical for limiting inflammation by terminating TNF-induced NF-kappa B responses.Specifica
| TNFAIP3 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP3 | |
| A20, OTUD7C, TNF alpha induced protein 3, TNFA1P2, TNFAIP3, Zinc finger protein A20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21929, 683206, 7128 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21580, Q60769 | |
| TNFAIP3 | |
| TNFAIP3 Fusion Protein Ag18150 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |