missing translation for 'onlineSavingsMsg'
Learn More

TNS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 20053-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899534

  • 129.00€ / 20µL

Spedizione stimata: 23-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The antibody recognizes all isoforms of TNS3.

The Tensin (Tns) family of proteins is involved in the maintece of cellular structure by anchoring actin filaments at the focal adhesion via F-actin binding and capping activities. Tns proteins also contain a Src homology 2 (SH2) domain and have the ability to be phosphorylated, suggesting a role in signal transduction cascades. These diverse characteristics indicate that Tns proteins may be important links between the cytoskeleton and signal transduction pathways. Tns3, also known as TEM6 or TENS1, localizes to the focal adhesions of the plasma membrane. It is predomitly expressed in thyroid and placenta but can also be found in heart, liver, brain, prostate, pancreas, kidney, lung, skeletal muscle and white blood cells. Tns3 is essential for proper growth and development, as suggested by growth retardation and death in a number of Tns3-/- mice.
Specifica

Specifica

TNS3
Polyclonal
Unconjugated
TNS3
H_NH049I23.2, TEM6, TENS1, tensin 3, TNS3, TPP, Tumor endothelial marker 6
Rabbit
Antigen Affinity Chromatography
RUO
319939, 360980, 64759
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5SSZ5, Q68CZ2
Tns3
Peptide
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali